Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnab1
Entrez Gene ID
16497
Gene Name
potassium voltage-gated channel, shaker-related subfamily, beta member 1
Gene Aliases
Akr8a8, Kvbeta1.1, mKv(beta)1
Chromosome Location
Chr.3: 64949049 - 65378225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289450.1 NP_001276379.1 9 - 10 594 61
NM_010597.4 NP_034727.3 8 - 9 1010 61
GenBank mRNA AF033003.1 - 8 - 9 1365 61
AK015412.1 - 10 - 11 850 61
AK031362.1 - 8 - 9 1011 61
AK138467.1 - 8 - 9 999 61
AK153200.1 - 9 - 10 596 61
BC014701.1 - 8 - 9 735 61
U65591.1 - 8 - 9 606 61
X97281.1 - 8 - 9 994 61

View Full Product Details