Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcnq2
Entrez Gene ID
16536
Gene Name
potassium voltage-gated channel, subfamily Q, member 2
Gene Aliases
HNSPC, KQT2, Nmf134
Chromosome Location
Chr.2: 181075579 - 181135289 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB000494.1 - 3 - 4 604 57
AB000495.1 - 3 - 4 604 57
AB000496.1 - 3 - 4 604 57
AB000497.1 - 3 - 4 604 57
AB000498.1 - 3 - 4 604 57
AB000499.1 - 3 - 4 604 57
AB000500.1 - 3 - 4 604 57
AB000501.1 - 3 - 4 604 57
AB000502.1 - 3 - 4 604 57
AB000503.1 - 3 - 4 604 57
AB000504.1 - 3 - 4 604 57
AF490773.1 - 3 - 4 518 57
AK135059.1 - 2 - 3 176 57
AK139411.1 - 3 - 4 349 57
BC145453.1 - 3 - 4 648 57

View Full Product Details