Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Meox1
Entrez Gene ID
17285
Gene Name
mesenchyme homeobox 1
Gene Aliases
AI385561, D330041M02Rik, Mox-1, Mox1
Chromosome Location
Chr.11: 101877510 - 101894354 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010791.3 NP_034921.1 1 - 2 813 95
GenBank mRNA AK052370.1 - 1 - 2 823 95
AK133672.1 - 1 - 2 184 95
BC011082.1 - 1 - 2 813 95
Z15103.1 - 1 - 2 768 95

View Full Product Details