The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122949.2 | NP_001116421.1 | 6 - 7 | 1022 | 94 |
NM_001285496.1 | NP_001272425.1 | 6 - 7 | 842 | 94 | |
NM_001285497.1 | NP_001272426.1 | 6 - 7 | 1022 | 94 | |
NM_010823.3 | NP_034953.2 | 6 - 7 | 998 | 94 | |
GenBank mRNA | AF360122.1 | - | 6 - 7 | 779 | 94 |
AK041166.1 | - | 6 - 7 | 970 | 94 | |
BC101943.1 | - | 6 - 7 | 979 | 94 | |
BC103513.1 | - | 6 - 7 | 955 | 94 | |
BC103514.1 | - | 6 - 7 | 979 | 94 | |
BC103515.1 | - | 6 - 7 | 979 | 94 | |
BC146612.1 | - | 6 - 7 | 951 | 94 | |
BC146613.1 | - | 6 - 7 | 956 | 94 | |
X73677.1 | - | 6 - 7 | 966 | 94 | |
Z22649.1 | - | 6 - 7 | 956 | 94 |