The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039657.2 | NP_001034746.1 | 3 - 4 | 755 | 75 |
NM_001039658.2 | NP_001034747.1 | 3 - 4 | 755 | 75 | |
NM_001286557.1 | NP_001273486.1 | 4 - 5 | 839 | 75 | |
NM_001286558.1 | NP_001273487.1 | 3 - 4 | 770 | 75 | |
GenBank mRNA | AB057423.1 | - | 3 - 4 | 743 | 75 |
AF329359.1 | - | 3 - 4 | 780 | 75 | |
AF329360.1 | - | 4 - 5 | 849 | 75 | |
AK015724.1 | - | 3 - 4 | 696 | 75 | |
AK015732.1 | - | 3 - 4 | 728 | 75 | |
AK161511.1 | - | 3 - 4 | 756 | 75 | |
BC024377.1 | - | 3 - 4 | 731 | 75 | |
U67176.1 | - | 5 - 6 | 772 | 75 | |
U77383.1 | - | 3 - 4 | 392 | 75 |