The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008681.2 | NP_032707.2 | 2 - 3 | 221 | 92 |
GenBank mRNA | AK004548.1 | - | 2 - 3 | 221 | 92 |
AK098075.1 | - | 2 - 3 | 220 | 92 | |
AK161860.1 | - | 2 - 3 | 223 | 92 | |
AK165007.1 | - | 2 - 3 | 221 | 92 | |
AK170281.1 | - | 2 - 3 | 222 | 92 | |
AK170439.1 | - | 2 - 3 | 220 | 92 | |
AK170636.1 | - | 2 - 3 | 220 | 92 | |
AK170809.1 | - | 2 - 3 | 221 | 92 | |
AK199494.1 | - | 1 - 2 | 45 | 92 | |
AW611038.1 | - | 2 - 3 | 221 | 92 | |
BC015282.1 | - | 2 - 3 | 176 | 92 | |
BC071235.1 | - | 2 - 3 | 229 | 92 | |
BC141246.1 | - | 2 - 3 | 112 | 92 | |
BC171968.1 | - | 2 - 3 | 112 | 92 | |
BY232155.1 | - | 2 - 3 | 222 | 92 | |
U52073.1 | - | 2 - 3 | 200 | 92 | |
U60593.1 | - | 2 - 3 | 219 | 92 |