Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk16
Entrez Gene ID
18555
Gene Name
cyclin-dependent kinase 16
Gene Aliases
Crk5, Pctaire1, Pctk1
Chromosome Location
Chr.X: 20687959 - 20699880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011049.4 NP_035179.1 15 - 16 1777 62
GenBank mRNA AK051425.1 - 15 - 16 1778 62
AK141083.1 - 9 - 10 797 62
AK143518.1 - 15 - 16 1805 62
AK145483.1 - 15 - 16 1807 62
AK166188.1 - 15 - 16 1986 62
BC011069.1 - 15 - 16 1520 62
BC013663.1 - 15 - 16 1836 62
X69025.1 - 15 - 16 1799 62

View Full Product Details