The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291434.1 | NP_001278363.1 | 3 - 4 | 543 | 89 |
NM_011102.4 | NP_035232.1 | 3 - 4 | 543 | 89 | |
GenBank mRNA | AK144419.1 | - | 3 - 4 | 545 | 89 |
BC111807.1 | - | 3 - 4 | 430 | 89 | |
L28035.1 | - | 3 - 4 | 621 | 89 | |
X67129.1 | - | 3 - 4 | 352 | 89 |