Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkcg
Entrez Gene ID
18752
Gene Name
protein kinase C, gamma
Gene Aliases
PKCgamma, Pkcc, Prkcc
Chromosome Location
Chr.7: 3303532 - 3331099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291434.1 NP_001278363.1 4 - 5 653 75
NM_011102.4 NP_035232.1 4 - 5 653 75
GenBank mRNA AK144419.1 - 4 - 5 655 75
BC111807.1 - 4 - 5 540 75
L28035.1 - 4 - 5 731 75
X67129.1 - 4 - 5 462 75

View Full Product Details