Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zp3
Entrez Gene ID
22788
Gene Name
zona pellucida glycoprotein 3
Gene Aliases
Zp-3
Chromosome Location
Chr.5: 135980105 - 135988624 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011776.1 NP_035906.1 3 - 4 558 72
GenBank mRNA AK131934.1 - 3 - 4 557 72
BC099465.1 - 3 - 4 543 72
BC100745.1 - 3 - 4 558 72
BC103583.1 - 3 - 4 558 72
BC103584.1 - 3 - 4 558 72
BC103585.1 - 3 - 4 558 72
M20026.1 - 3 - 4 558 72

View Full Product Details