Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zp3
Entrez Gene ID
22788
Gene Name
zona pellucida glycoprotein 3
Gene Aliases
Zp-3
Chromosome Location
Chr.5: 135980105 - 135988624 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011776.1 NP_035906.1 5 - 6 859 83
GenBank mRNA AK131934.1 - 5 - 6 858 83
BC099465.1 - 5 - 6 844 83
BC100745.1 - 5 - 6 859 83
BC103583.1 - 5 - 6 859 83
BC103584.1 - 5 - 6 859 83
BC103585.1 - 5 - 6 859 83
M20026.1 - 5 - 6 859 83

View Full Product Details