The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038018.4 | NP_001033107.1 | 12 - 13 | 1439 | 68 |
NM_001112711.2 | NP_001106182.1 | 12 - 13 | 1439 | 68 | |
NM_001286063.1 | NP_001272992.1 | 12 - 13 | 1419 | 68 | |
NM_001286064.1 | NP_001272993.1 | 12 - 13 | 1424 | 68 | |
NM_001286065.1 | NP_001272994.1 | 12 - 13 | 1337 | 68 | |
NM_001286066.1 | NP_001272995.1 | 12 - 13 | 1432 | 68 | |
NM_011938.4 | NP_036068.2 | 12 - 13 | 1439 | 68 | |
GenBank mRNA | AF040747.1 | - | 12 - 13 | 1309 | 68 |
AF040748.1 | - | 12 - 13 | 1309 | 68 | |
AF040749.1 | - | 12 - 13 | 1309 | 68 | |
AK051405.1 | - | 12 - 13 | 1421 | 68 | |
AK146119.1 | - | 12 - 13 | 1212 | 68 | |
AK161142.1 | - | 10 - 11 | 1071 | 68 | |
BC057206.1 | - | 12 - 13 | 1413 | 68 | |
BC075719.1 | - | 12 - 13 | 1320 | 68 | |
CA528447.1 | - | 1 - 2 | 38 | 68 | |
Y15797.1 | - | 12 - 13 | 1261 | 68 | |
Y15798.1 | - | 12 - 13 | 1283 | 68 | |
Y15799.1 | - | 12 - 13 | 1266 | 68 | |
Y15800.1 | - | 12 - 13 | 1254 | 68 |