The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013743.2 | NP_038771.1 | 6 - 7 | 812 | 78 |
GenBank mRNA | AJ001418.1 | - | 6 - 7 | 812 | 78 |
AK004543.1 | - | 6 - 7 | 837 | 78 | |
AK036818.1 | - | 6 - 7 | 844 | 78 | |
BC026134.1 | - | 6 - 7 | 841 | 78 |