Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hunk
Entrez Gene ID
26559
Gene Name
hormonally upregulated Neu-associated kinase
Gene Aliases
Bstk1, Mak-v
Chromosome Location
Chr.16: 90385797 - 90499555 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015755.2 NP_056570.1 10 - 11 1541 89
GenBank mRNA AF055919.2 - 10 - 11 1541 89
AF167987.1 - 10 - 11 1553 89
AK144639.1 - 10 - 11 1524 89
AK147425.1 - 10 - 11 1927 89
AK147734.1 - 10 - 11 1295 89
BC137788.1 - 10 - 11 1602 89
BC137790.1 - 10 - 11 1601 89

View Full Product Details