Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dkk3
Entrez Gene ID
50781
Gene Name
dickkopf homolog 3 (Xenopus laevis)
Gene Aliases
AW061014, C87148
Chromosome Location
Chr.7: 112116019 - 112159121 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015814.2 NP_056629.1 4 - 5 543 96
GenBank mRNA AF177400.1 - 4 - 5 530 96
AJ243964.1 - 4 - 5 546 96
AK004853.1 - 4 - 5 543 96
AK013054.2 - 3 - 4 481 96
AK013622.1 - 3 - 4 455 96
BC046304.1 - 3 - 4 458 96
BC050934.1 - 3 - 4 440 96

View Full Product Details