Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chek2
Entrez Gene ID
50883
Gene Name
checkpoint kinase 2
Gene Aliases
CHK2, Cds1, HUCDS1, Rad53
Chromosome Location
Chr.5: 110839791 - 110874140 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016681.3 NP_057890.1 3 - 4 496 91
GenBank mRNA AF086905.1 - 2 - 3 457 91
AK044913.1 - 3 - 4 497 91
AK170235.1 - 3 - 4 531 91
BC056617.1 - 3 - 4 520 91

View Full Product Details