Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtx2
Entrez Gene ID
53375
Gene Name
metaxin 2
Gene Aliases
1500012G02Rik
Chromosome Location
Chr.2: 74825812 - 74876748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016804.4 NP_058084.3 1 - 2 236 70
GenBank mRNA AB041562.1 - 1 - 2 228 70
AF053550.1 - 1 - 2 209 70
AK005233.1 - 1 - 2 168 69
AK083055.1 - - 198 70
AK168000.1 - 1 - 2 229 70
AK168021.1 - 1 - 2 199 70
AK168045.1 - 1 - 2 228 70
AK169048.1 - 1 - 2 232 70
BC006641.1 - 1 - 2 186 70
BY126313.1 - 1 - 2 245 70

View Full Product Details