Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgl1
Entrez Gene ID
19731
Gene Name
ral guanine nucleotide dissociation stimulator,-like 1
Gene Aliases
Rgl, mKIAA0959
Chromosome Location
Chr.1: 152516768 - 152766351 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016846.3 NP_058542.2 17 - 18 2215 71
GenBank mRNA AK144218.1 - 17 - 18 2290 71
AK169109.1 - 17 - 18 2267 71
AK169180.1 - 17 - 18 2268 71
AK173071.1 - 7 - 8 810 71
BC017678.1 - 17 - 18 2215 71
U14103.1 - 17 - 18 2273 71

View Full Product Details