The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301692.1 | NP_001288621.1 | 5 - 6 | 703 | 75 |
NM_001301694.1 | NP_001288623.1 | 5 - 6 | 653 | 75 | |
NM_021340.4 | NP_067315.1 | 6 - 7 | 825 | 75 | |
GenBank mRNA | AF076930.1 | - | 6 - 7 | 745 | 75 |
AK044450.1 | - | 6 - 7 | 777 | 75 | |
AK044649.1 | - | 6 - 7 | 776 | 75 | |
AK149272.1 | - | 6 - 7 | 776 | 75 | |
BC082611.1 | - | 6 - 7 | 762 | 75 | |
BC119055.1 | - | 6 - 7 | 759 | 75 | |
BC144937.1 | - | 5 - 6 | 638 | 75 | |
CK623457.1 | - | 5 - 6 | 584 | 75 |