The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021424.2 | NP_067399.2 | 5 - 6 | 1072 | 92 |
GenBank mRNA | AF239762.1 | - | 5 - 6 | 1004 | 92 |
AF297665.1 | - | 5 - 6 | 1019 | 92 | |
AK147479.1 | - | - | 1664 | 92 | |
BC060694.1 | - | 5 - 6 | 1072 | 92 |