The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021600.3 | NP_067611.2 | 6 - 7 | 719 | 68 |
GenBank mRNA | AK081344.1 | - | 5 - 6 | 705 | 68 |
BC052153.1 | - | 6 - 7 | 664 | 68 | |
BC141376.1 | - | 6 - 7 | 647 | 68 | |
BC141387.1 | - | 6 - 7 | 647 | 68 | |
K02582.1 | - | 6 - 7 | 630 | 68 | |
L10076.1 | - | 6 - 7 | 679 | 68 |