The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159769.2 | NP_001153241.1 | 2 - 3 | 201 | 104 |
NM_030676.3 | NP_109601.1 | 2 - 3 | 286 | 104 | |
GenBank mRNA | AB221629.1 | - | 2 - 3 | 128 | 104 |
BB652042.1 | - | 2 - 3 | 172 | 104 | |
BC137845.1 | - | 2 - 3 | 165 | 104 | |
DQ133942.1 | - | 2 - 3 | 201 | 104 | |
M81385.1 | - | 2 - 3 | 286 | 104 |