The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159482.1 | NP_001152954.1 | 10 - 11 | 954 | 68 |
NM_033475.3 | NP_258436.2 | 9 - 10 | 1299 | 68 | |
GenBank mRNA | AB082927.1 | - | 9 - 10 | 1212 | 68 |
AB232630.1 | - | 9 - 10 | 710 | 68 | |
AF322068.1 | - | 9 - 10 | 710 | 68 | |
AF327929.1 | - | 10 - 11 | 904 | 68 | |
AK160308.1 | - | - | 1268 | 68 | |
AK181473.1 | - | 2 - 3 | 124 | 68 | |
BC038638.1 | - | 9 - 10 | 774 | 68 | |
S72304.1 | - | 7 - 8 | 557 | 68 |