The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161667.1 | NP_001155139.1 | 5 - 6 | 729 | 69 |
NM_053115.2 | NP_444345.2 | 5 - 6 | 854 | 69 | |
GenBank mRNA | AJ238492.1 | - | 5 - 6 | 771 | 69 |
BC021339.1 | - | 5 - 6 | 733 | 69 |