The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161667.1 | NP_001155139.1 | 14 - 15 | 2130 | 99 |
NM_053115.2 | NP_444345.2 | 14 - 15 | 2255 | 99 | |
GenBank mRNA | AJ238492.1 | - | 14 - 15 | 2172 | 99 |
AK144243.1 | - | 14 - 15 | 2124 | 99 | |
BC021339.1 | - | 14 - 15 | 2134 | 99 |