The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080466.2 | NP_536714.2 | 2 - 3 | 1334 | 71 |
GenBank mRNA | AF357241.1 | - | 2 - 3 | 1074 | 71 |
AK137738.1 | - | 2 - 3 | 1335 | 71 | |
BC106769.1 | - | 2 - 3 | 1076 | 71 | |
BC106770.1 | - | 2 - 3 | 1076 | 71 |