The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133184.2 | NP_573447.2 | 5 - 6 | 558 | 88 |
GenBank mRNA | AF251267.1 | - | 5 - 6 | 558 | 88 |