The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037127.2 | NP_001032204.2 | 14 - 15 | 2003 | 122 |
NM_001037128.1 | NP_001032205.1 | 14 - 15 | 1958 | 122 | |
NM_001037129.1 | NP_001032206.1 | 13 - 14 | 1934 | 122 | |
NM_001037130.1 | NP_001032207.1 | 12 - 13 | 1904 | 122 | |
NM_010944.2 | NP_035074.2 | 13 - 14 | 1928 | 122 | |
GenBank mRNA | AB691772.1 | - | 12 - 13 | 1754 | 122 |
AY955296.1 | - | 14 - 15 | 1853 | 122 | |
AY955297.1 | - | 14 - 15 | 1808 | 122 | |
AY955298.1 | - | 13 - 14 | 1784 | 122 | |
AY956403.1 | - | 13 - 14 | 1778 | 122 | |
AY956404.1 | - | 12 - 13 | 1754 | 122 | |
BC100342.1 | - | 13 - 14 | 1858 | 122 | |
U37708.1 | - | 12 - 13 | 1902 | 122 | |
U37709.1 | - | 13 - 14 | 1926 | 122 | |
X86444.1 | - | 12 - 13 | 1832 | 122 |