Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Slc34a2
- Entrez Gene ID
-
20531
- Gene Name
- solute carrier family 34 (sodium phosphate), member 2
- Gene Aliases
-
AA536683, D5Ertd227e, NaPi-2b, Npt2b
- Chromosome Location
-
Chr.5: 53049203 - 53071664 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF081499.1 |
- |
13 - 14 |
2939 |
90 |
View Full Product Details