The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011545.1 | NP_035675.1 | 1 - 2 | 677 | 68 |
GenBank mRNA | AB009453.1 | - | 1 - 2 | 674 | 68 |
AF029753.1 | - | 1 - 2 | 664 | 68 | |
AF035717.1 | - | 1 - 2 | 677 | 68 | |
AF036945.1 | - | 1 - 2 | 715 | 68 | |
AF047418.1 | - | 1 - 2 | 650 | 68 | |
AK011575.1 | - | 1 - 2 | 692 | 68 | |
AK135423.1 | - | 1 - 2 | 724 | 68 | |
AK157289.1 | - | 1 - 2 | 693 | 68 | |
BC053525.1 | - | 1 - 2 | 684 | 68 |