The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011904.3 | NP_036034.1 | 1 - 2 | 297 | 139 |
GenBank mRNA | AF073526.1 | - | 1 - 2 | 358 | 139 |
BC132537.1 | - | 1 - 2 | 297 | 139 | |
BC145132.1 | - | 1 - 2 | 297 | 139 |