The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110780.1 | NP_001104250.1 | 10 - 11 | 1434 | 78 |
NM_013680.4 | NP_038708.3 | 10 - 11 | 1434 | 78 | |
GenBank mRNA | BC022954.1 | - | 10 - 11 | 1412 | 78 |