The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017461.2 | NP_059489.2 | 6 - 7 | 594 | 111 |
GenBank mRNA | BB610735.1 | - | 6 - 7 | 594 | 111 |
BC140998.1 | - | 6 - 7 | 595 | 111 | |
BC145081.1 | - | 6 - 7 | 595 | 111 | |
M37030.1 | - | 6 - 7 | 598 | 111 |