Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex3
Entrez Gene ID
56535
Gene Name
peroxisomal biogenesis factor 3
Gene Aliases
1700014F15Rik, 2810027F19Rik, 2900010N04Rik
Chromosome Location
Chr.10: 13523842 - 13553149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164195.1 NP_001157667.1 10 - 11 1183 85
NM_019961.3 NP_064345.1 10 - 11 1183 85
GenBank mRNA AF152996.1 - 10 - 11 1106 85
AK005315.1 - 10 - 11 1039 85
AK005978.1 - 9 - 10 880 85
AK012827.1 - 10 - 11 1161 85
AK075820.1 - 10 - 11 1098 85
AK146934.1 - 10 - 11 1162 85
BC033415.1 - 10 - 11 1065 85
BC037606.1 - 10 - 11 1146 85

View Full Product Details