The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019969.3 | NP_064353.2 | 5 - 6 | 839 | 72 |
GenBank mRNA | AF057366.1 | - | 1 - 2 | 256 | 72 |
AY574219.1 | - | 5 - 6 | 816 | 72 | |
BC139283.1 | - | 5 - 6 | 789 | 72 | |
BC139285.1 | - | 5 - 6 | 789 | 72 |