The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004468.2 | NP_001004468.1 | 21 - 22 | 8574 | 80 |
NM_021314.4 | NP_067289.2 | 17 - 18 | 3409 | 80 | |
NM_206856.3 | NP_996738.2 | 20 - 21 | 3375 | 80 | |
GenBank mRNA | AB041546.1 | - | 17 - 18 | 3407 | 80 |
AK145721.1 | - | 15 - 16 | 3038 | 80 | |
AK165194.1 | - | 16 - 17 | 3233 | 80 | |
AK165603.1 | - | 14 - 15 | 2933 | 80 | |
AK220527.1 | - | 3 - 4 | 3212 | 80 | |
AY177410.1 | - | 19 - 20 | 3384 | 80 | |
BC004057.1 | - | 12 - 13 | 2449 | 80 | |
BC109016.1 | - | 4 - 5 | 3467 | 80 |