The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021407.3 | NP_067382.1 | 2 - 3 | 515 | 70 |
GenBank mRNA | AF241220.1 | - | 2 - 3 | 467 | 70 |
AK079407.1 | - | 2 - 3 | 440 | 70 | |
BC127035.1 | - | 2 - 3 | 450 | 70 | |
BC127036.1 | - | 2 - 3 | 450 | 70 |