Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mkks
Entrez Gene ID
59030
Gene Name
McKusick-Kaufman syndrome
Gene Aliases
1300013E18Rik, AI463362, AI957237, Bbs6
Chromosome Location
Chr.2: 136873780 - 136891505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286981.1 NP_001273910.1 1 - 2 364 90
NM_001286983.1 NP_001273912.1 1 - 2 364 90
NM_021527.2 NP_067502.2 1 - 2 364 90
GenBank mRNA AF254074.1 - 1 - 2 136 90
AK004987.1 - 1 - 2 121 90
AK008160.1 - 1 - 2 106 90
AK008406.1 - 1 - 2 106 90
AK032528.1 - 1 - 2 126 90
AK032554.1 - 1 - 2 126 90
BC058174.1 - 1 - 2 347 90
BC100336.1 - 1 - 2 117 90

View Full Product Details