The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021883.2 | NP_068683.1 | 6 - 7 | 755 | 74 |
GenBank mRNA | AK132878.1 | - | 5 - 6 | 588 | 74 |
BC106849.1 | - | 5 - 6 | 643 | 74 | |
BC106850.1 | - | 5 - 6 | 643 | 74 | |
S76831.1 | - | 5 - 6 | 682 | 74 |