The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024211.3 | NP_077173.1 | 1 - 2 | 268 | 97 |
GenBank mRNA | AI931249.1 | - | 1 - 2 | 229 | 97 |
AK009487.1 | - | 1 - 2 | 214 | 97 | |
AK009824.1 | - | 1 - 2 | 168 | 97 | |
AK168951.1 | - | 1 - 2 | 193 | 97 | |
BC003455.1 | - | 1 - 2 | 244 | 97 | |
BC019631.1 | - | 1 - 2 | 222 | 97 |