The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289603.1 | NP_001276532.1 | 1 - 2 | 99 | 52 |
NM_033476.3 | NP_258437.1 | 1 - 2 | 99 | 52 | |
GenBank mRNA | AK146503.1 | - | 1 - 2 | 100 | 52 |
BC022131.1 | - | 1 - 2 | 72 | 52 | |
M84809.1 | - | 1 - 2 | 38 | 52 |