The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289603.1 | NP_001276532.1 | 4 - 5 | 488 | 69 |
NM_033476.3 | NP_258437.1 | 4 - 5 | 488 | 69 | |
GenBank mRNA | AK131989.1 | - | 3 - 4 | 687 | 69 |
AK146503.1 | - | 4 - 5 | 489 | 69 | |
AK147898.1 | - | 3 - 4 | 697 | 69 | |
AK158682.1 | - | 4 - 5 | 408 | 69 | |
BC022131.1 | - | 4 - 5 | 461 | 69 | |
M84809.1 | - | 4 - 5 | 427 | 69 |