Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfcp2
Entrez Gene ID
21422
Gene Name
transcription factor CP2
Gene Aliases
CP-2, CP2, D230015P20Rik, LBP-1c, LBP-1d, LBP1, LSF, Tcfcp2, UBP-1
Chromosome Location
Chr.15: 100502748 - 100552014 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289603.1 NP_001276532.1 14 - 15 1549 80
NM_033476.3 NP_258437.1 14 - 15 1555 80
GenBank mRNA AK013652.1 - 1 - 2 69 80
AK141248.1 - 8 - 9 895 80
AK146503.1 - 14 - 15 1556 80
AK158682.1 - 14 - 15 1475 80
BC022131.1 - 14 - 15 1522 80
M84809.1 - 14 - 15 1494 80

View Full Product Details