The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290651.1 | NP_001277580.1 | 25 - 26 | 3227 | 90 |
NM_001290652.1 | NP_001277581.1 | 23 - 24 | 3047 | 90 | |
GenBank mRNA | AF285577.1 | - | 22 - 23 | 2633 | 90 |
AK016533.1 | - | 22 - 23 | 2596 | 90 | |
AK031490.1 | - | 25 - 26 | 3208 | 90 | |
AK040521.1 | - | 23 - 24 | 3049 | 90 |