The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139310.1 | NP_647471.1 | 15 - 16 | 1703 | 85 |
GenBank mRNA | AK157904.1 | - | 15 - 16 | 1735 | 85 |
AK158092.1 | - | 15 - 16 | 1818 | 85 | |
AK158280.1 | - | 5 - 6 | 676 | 85 | |
AK158326.1 | - | 5 - 6 | 672 | 85 | |
AY055122.1 | - | 15 - 16 | 1703 | 85 |