The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147201.2 | NP_671734.1 | 14 - 15 | 1438 | 56 |
GenBank mRNA | AF302138.1 | - | 14 - 15 | 1437 | 56 |
AK046142.1 | - | 15 - 16 | 1456 | 56 | |
AK077440.1 | - | 14 - 15 | 1475 | 56 | |
AK170501.1 | - | 14 - 15 | 1454 | 56 | |
AK185308.1 | - | 3 - 4 | 225 | 56 | |
AK196002.1 | - | 3 - 4 | 225 | 56 | |
AK215481.1 | - | 3 - 4 | 284 | 56 | |
BC004756.1 | - | 14 - 15 | 1408 | 56 | |
BC013212.1 | - | 3 - 4 | 832 | 56 | |
BC018463.1 | - | 14 - 15 | 1412 | 56 |