The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_148933.1 | NP_683735.1 | 1 - 2 | 221 | 62 |
GenBank mRNA | BC030719.1 | - | 1 - 2 | 221 | 62 |
BC030720.1 | - | 1 - 2 | 170 | 62 |