The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008695.2 | NP_032721.2 | 13 - 14 | 3195 | 87 |
GenBank mRNA | AB017202.1 | - | 13 - 14 | 3157 | 87 |
AK045306.1 | - | 13 - 14 | 3170 | 87 | |
AK051664.1 | - | 13 - 14 | 3171 | 87 | |
AK052839.1 | - | 13 - 14 | 3169 | 87 | |
AK083629.1 | - | 13 - 14 | 87 | ||
AK128978.1 | - | 10 - 11 | 2317 | 87 | |
AK140832.1 | - | 13 - 14 | 3169 | 87 | |
AK164258.1 | - | 13 - 14 | 3166 | 87 | |
AY074820.1 | - | 13 - 14 | 3156 | 87 | |
BC054746.1 | - | 13 - 14 | 3177 | 87 | |
BC057016.1 | - | 13 - 14 | 3123 | 87 |