Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St8sia2
Entrez Gene ID
20450
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
Gene Aliases
AI323367, ST8SiaII, STX, Siat8b
Chromosome Location
Chr.7: 73939119 - 74013727 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009181.2 NP_033207.2 1 - 2 289 80
GenBank mRNA BC096546.1 - 1 - 2 281 80
X83562.1 - 1 - 2 268 80

View Full Product Details