Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppp2r5d
Entrez Gene ID
21770
Gene Name
protein phosphatase 2, regulatory subunit B', delta
Gene Aliases
B'delta, TEG-271, Tex271
Chromosome Location
Chr.17: 46682991 - 46705020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009358.3 NP_033384.2 14 - 15 1797 68
GenBank mRNA AB053464.1 - 14 - 15 1742 68
AF316616.2 - 14 - 15 1654 68
AK154050.1 - 14 - 15 1773 68
AK160587.1 - 14 - 15 1738 68
AK178339.1 - 1 - 2 139 68
AY338234.1 - 14 - 15 1727 68
BC010716.1 - 14 - 15 1710 68
X81059.1 - 3 - 4 237 68

View Full Product Details